missing translation for 'onlineSavingsMsg'
Learn More

GSK319347A, MedChemExpress

Product Code. 16428737
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16428737 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16428737 Supplier Medchem Express Supplier No. HY14682
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress GSK319347A is a dual inhibitor of TBK1 and IKKε with IC50s of 93 nM and 469 nM, respectively. GSK319347A also inhibits IKK2 with an IC50 of 790 nM.

GSK319347A is a dual inhibitor of TBK1 and IKKε with IC50s of 93 nM and 469 nM, respectively. GSK319347A also inhibits IKK2 with an IC50 of 790 nM.

Specifications

Chemical Name or Material GSK319347A
CAS 862812-98-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H19N3O5S2
Quantity 5 mg
Solubility Information DMSO : 45 mg/mL (95.84 mM; Need ultrasonic)
SMILES COC1=CC2=C(C=C1OC)N(C3=CC(OCC4=CC=CC=C4S(=O)(C)=O)=C(C#N)S3)C=N2
Molecular Weight (g/mol) 469.53
Formula Weight 469.53
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.