missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ GSK583, 95%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | GSK583 |
| CAS | 1346547-00-9 |
| Synonym | 6-(tert-Butylsulfonyl)-N-(5-fluoro-1 H-indazol-3-yl)quinolin-4-amine |
| Quantity | 100 mg |
| Molecular Formula | C20 H19 F N4 O2 S |
| Formula Weight | 398.45 |
| Physical Form | Solid |
| Product Type | Reagent |
| Purity | 95% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction