missing translation for 'onlineSavingsMsg'
Learn More

Guaiazulene, 99%

Product Code. 11984451 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11984451 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11984451 Supplier Thermo Scientific Acros Supplier No. 120200100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 489-84-9 | C15H18 | 198.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 489-84-9
Molecular Formula C15H18
Molecular Weight (g/mol) 198.31
InChI Key FWKQNCXZGNBPFD-UHFFFAOYSA-N
Synonym guaiazulene, 1,4-dimethyl-7-isopropylazulene, azulon, vetivazulen, azunol, 7-isopropyl-1,4-dimethylazulene, eucazulen, guajazulene, kessazulen, purazulen
PubChem CID 3515
ChEBI CHEBI:5550
IUPAC Name 1,4-dimethyl-7-propan-2-ylazulene
SMILES CC1=C2C=CC(=C2C=C(C=C1)C(C)C)C

Spécification

Melting Point 29.0°C to 31.0°C
Color Blue
Density 0.9700g/mL
Boiling Point 153.0°C (7.0 Torr)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 05,II,473
Merck Index 15,459
Specific Gravity 0.97
Solubility Information Solubility in water: 25%. Other solubilities: soluble in ethanol, ethyl ether and ethyl acetate
Formula Weight 198.31
Percent Purity 99%
Physical Form Low Melting Crystalline Solid
Chemical Name or Material Guaiazulene
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • Guaiazulene
Mention d'avertissement
  • Warning
Catégorie de danger
  • Acute toxicity Category 4
Mention de danger
  • H302-Harmful if swallowed.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.