missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Guanfacine hydrochloride

Product Code. 30001167
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001167 50 mg 50mg
18742624 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001167 Supplier R&D Systems™ Supplier No. 1030/50
  • ONLINE EXCLUSIVE PRICE
    435.85 €
    392.70 € / 50mg
    Save 43.15 €
    9% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. α2A agonist

Guanfacine hydrochloride is a selective α2A-adrenoceptor agonist (Kd = 31 nM). Displays 60-fold selectivity over α2B-adrenoceptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29110-48-3
Molecular Formula C9H10Cl3N3O

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 282.56
Purity >99%
Molecular Weight (g/mol) 282.549
Chemical Name or Material Guanfacine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.