missing translation for 'onlineSavingsMsg'
Learn More

Guanidine carbonate, 98%

Product Code. 11465263
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465263 5000 g 5000g
11455263 250 g 250g
11445263 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11465263 Supplier Thermo Scientific Alfa Aesar Supplier No. A12853.0I
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 593-85-1 | C3H12N6O3

It is employed as a strong organic alkali, organic intermediate, in soap and cosmetic products, and in the textile industry. It is also used to make fine chemicals. It is used in various industries to solve different purpose such as detergents as a synergistic agent, pharmaceuticals as an antibiotic and microencapsulation process for carbonless paper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a strong organic alkali, organic intermediate, in soap and cosmetic products, and in the textile industry. It is also used to make fine chemicals. It is used in various industries to solve different purpose such as detergents as a synergistic agent, pharmaceuticals as an antibiotic and microencapsulation process for carbonless paper.

Solubility
Soluble in water 450 g/L (20°C); slightly soluble in alcohol and acetone.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-85-1
Molecular Formula C3H12N6O3
MDL Number MFCD00013029

Specifications

Melting Point ∼198°C (decomposition)
Quantity 5000 g
Beilstein 3628359
Solubility Information Soluble in water 450g/L (20°C); slightly soluble in alcohol and acetone.
Formula Weight 180.17
Percent Purity 98%
Chemical Name or Material Guanidine carbonate
compliance-icons
Product Identifier
  • Guanidine carbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Guanidine carbonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.