missing translation for 'onlineSavingsMsg'
Learn More

Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2)

Product Code. 10132340 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
10 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
250g
25kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10132340 250 g Plastic bottle 250g
10447690 1 kg Plastic bottle 1kg
10337180 2.5 kg Plastic bottle 2.5kg
10573174 10 kg Plastic drum 10kg
11965591 25 kg Fibre drum 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10132340 Supplier Thermo Scientific Acros Supplier No. 120232500
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2), Quantity: 1kg, Packaging: Plastic bottle, White to Yellow, Melting Point: 179.0 deg.C to 186.0 deg.C, Molecular Weight: 95.53, Percent Purity: 98%, Assay Percent Range: 0.5% max. Ammonium chloride (Argentometry) | CAS: 50-01-1 | CH5N3·HCl | 95.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH5N3·HCl
Molecular Weight (g/mol) 95.53
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl

Specifications

Melting Point 179.0°C to 186.0°C
Color White to Yellow
Density 1.3400g/mL
Infrared Spectrum Authentic
Assay Percent Range 0.5% max. Ammonium chloride (Argentometry)
Packaging Plastic bottle
Linear Formula H2NC(=NH)NH2·HCl
Quantity 250 g
Beilstein 03,82
Merck Index 15,4597
Specific Gravity 1.34
Solubility Information Solubility in water: 2280g/L (20°C). Other solubilities: 300g/L ethanol (30°C), 19.5g/100g dimethylformamide (30°C), <0.05g/100g acetone -<0.05g/100g benzene (30°C), insoluble in trichloroethylene
Formula Weight 95.53
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Guanidine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Guanidine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.