missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Hydrochloride, >99.5%, Molecular Biology Grade

Product Code. 15845228 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Poly pail
Unit Size:
100g
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 15845228

Brand: Thermo Scientific J75823.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50-01-1 | CH6ClN3 | 95.53 g/mol

  • Used for extraction of RNA.
  • Preparation of high molecular weight genomic DNA.
  • Solubilizing inclusion bodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Used for extraction of RNA
  • Preparation of high molecular weight genomic DNA
  • Solubilizing inclusion bodies
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
MDL Number MFCD00013026
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl

Specifications

Melting Point 185°C to 188°C
Density 1.344
Packaging Plastic bottle
Linear Formula (NH2)2C=NH·HCl
Quantity 500 g
Beilstein 3591990
Formula Weight 95.53
Percent Purity ≥99.5%
Grade Molecular Biology
Physical Form Crystals
Chemical Name or Material Guanidine Hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Guanidine hydrochloride, Molecular Biology Grade, Thermo Scientific
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Guanidine, hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.