missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Hydrochloride, TRC

Product Code. 30133207
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133207 25 g 25g
30011498 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133207 Supplier Toronto Research Chemicals Supplier No. TRCG82124525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-01-1
Molecular Formula C H5 N3 . Cl H
Molecular Weight (g/mol) 95.53
Synonym Guanidine, hydrochloride (1:1), Guanidine, monohydrochloride (8CI,9CI), Buffer AL, Guanidine chloride, Guanidine hydrochloride, Guanidinium chloride, Guanidinium hydrochloride
IUPAC Name guanidine;hydrochloride
SMILES Cl.NC(=N)N

Specifications

Quantity 25 g
InChI Formula InChI=1S/CH5N3.ClH/c2-1(3)4;/h(H5,2,3,4);1H
Formula Weight 95.025
Chemical Name or Material Guanidine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.