missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Hydrochloride, Ultra Pure, 500g, MP Biomedicals

Product Code. 11451682 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11451682 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11451682 Supplier MP Biomedicals Supplier No. 04820539
  • ONLINE EXCLUSIVE PRICE
    405.00 €
    383.25 € / 500g
    Save 21.75 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

50-01-1 Purity: >>99% O.D. (260 nm) of a 6 M solution: <<0.03 Ammeline: none detected Protein denaturant for molecular weight determinations

Specifications

CAS 50-01-1
Molecular Formula CH6ClN3
Quantity 500 g
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
SMILES C(=N)(N)N.Cl
IUPAC Name guanidine;hydrochloride
Molecular Weight (g/mol) 95.53
PubChem CID 5742
ChEBI CHEBI:32735
Grade Ultrapure
Chemical Name or Material Guanidine Hydrochloride, Ultra Pure
Show More Show Less
compliance-icons
Product Identifier
  • Guanidine Hydrochloride, ultra pure
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P235-Keep cool.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: guanidine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.