missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Hydrochloride, Ultra Pure grade, MP Biomedicals

Product Code. 11441682 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441682 100 g 100g
11461682 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441682 Supplier MP Biomedicals Supplier No. 04820512
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

  • Guanidine Hydrochloride is a strong chaotropic agent useful for the denaturation and subsequent refolding of proteins
  • This strong denaturant can solubilize insoluble or denatured proteins such as inclusion bodies
  • This can be used as the first step in refolding proteins or enzymes into their active form
  • Urea and dithiothreitol (DTT) may also be necessary
  • Guanidine HCl is used in the isolation of RNA to dissociate the nucleoprotein into its nucleic acid and protein moieties
  • It is an inhibitor of RNase
  • Highly concentrated (6 - 8 M) Guanidine HCl solutions are used to denature native globular proteins
  • It apparently disrupts hydrogen bonds which hold the protein in its unique structure; however, there also is evidence suggesting that guanidine hydrocholoride may disrupt hydrophobic interactions by promoting the solubility of hydrophobic residues in aqueous solutions

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
MDL Number MFCD00013026
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl

Specifications

Melting Point 180°C to 190°C
Color White
Linear Formula CH5N3·HCl
Quantity 100 g
Solubility Information Soluble in water (6 M - clear, colorless solution).
Formula Weight 95.53
Percent Purity ≥99%
Grade Ultra Pure
Physical Form Crystalline Powder
Chemical Name or Material GUANIDINE HYDROCHLORIDE
compliance-icons
Product Identifier
  • Guanidine Hydrochloride, ultra pure
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: guanadine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.