missing translation for 'onlineSavingsMsg'
Learn More

Guanidine nitrate, 98%, Thermo Scientific Chemicals

Product Code. 11342177
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1kg
250g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11342177

Brand: Thermo Scientific Alfa Aesar L09643.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-93-4
Molecular Formula CH6N4O3
Molecular Weight (g/mol) 122.084
MDL Number MFCD00013028
InChI Key CNUNWZZSUJPAHX-UHFFFAOYSA-N
Synonym guanidine nitrate, guanidinium nitrate, guanidine; nitric acid, guanidine, mononitrate, guanidine, nitrate, guanidine mononitrate, guanidine nitrate 1:1, guanidinenitrate, pubchem21372, acmc-209kpa
PubChem CID 10481
IUPAC Name guanidine;nitric acid
SMILES C(=N)(N)N.[N+](=O)(O)[O-]

Specifications

Melting Point 213°C to 216°C
Density 1.44
pH 4.9
Odor Odorless
Quantity 250 g
UN Number UN1467
Beilstein 3596600
Merck Index 14,4562
Formula Weight 122.08
Percent Purity 98%
Chemical Name or Material Guanidine nitrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.