missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Nitrate, TRC

Product Code. 30393160
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30393160 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30393160

Brand: Toronto Research Chemicals TRCG82124010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Guanidine Nitrate
CAS 506-93-4
Molecular Formula CH5N4O3
Quantity 10 g
Synonym Guanidinium Nitrate, NSC 7295, Guanidine Mononitrate
InChI Formula InChI=1S/CH5N3.HNO3/c2*2-1(3)4/h(H5,2,3,4);(H,2,3,4)
SMILES NC(=N)N.O[N+](=O)[O-]
IUPAC Name guanidine;nitric acid
Molecular Weight (g/mol) 121.08
Formula Weight 122.044
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.