missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Thiocyanate, MP Biomedicals™

Product Code. 11491682 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491682 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11491682 Supplier MP Biomedicals Supplier No. 04820991
  • ONLINE EXCLUSIVE PRICE
    319.00 €
    302.40 € / 500g
    Save 16.60 €
    5% Off
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Guanidine Thiocyanate, Research Grade, >98%, Hard, White Crystalline Granules

  • Guanidine Thiocyanate is often used in the isolation of intact ribonucleic acid to eliminate RNase activity.
  • A 4 M solution of guanidine thiocyanate irreversibly inactivates RNase.
  • In the presence of guanidine thiocyanate, proteins dissolve readily, cellular structures disintegrate and nucleoproteins dissociate from nucleic acids, as protein secondary structure is lost.
  • Guanidine Thiocyanate is a chaotropic agent and strong protein denaturant which inactivates nucleases approximately 2.5 times faster than guanidine hydrochloride.

Specifications

CAS 593-84-0
Melting Point 115°C to 122°C
Color White
pH 5 to 7 (6M aq soln)
Loss on Drying <1%
Molecular Formula C2H6N4S
MDL Number MFCD00013027
Quantity 500 g
UN Number UN1759
Beilstein 3563461
Synonym Guanidinium rhodanide, Guanidinium thiocyanate
Solubility Information Soluble in (6.0 M aq soln): Clear, Colorless Solution.
InChI Key ZJYYHGLJYGJLLN-UHFFFAOYSA-N
Molecular Weight (g/mol) 118.16
Grade >98%
Physical Form Crystalline Granules
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.