missing translation for 'onlineSavingsMsg'
Learn More

Guanine, 99+%

Product Code. 10255060 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10255060 10 g Glass bottle 10g
10697262 25 g Glass bottle 25g
10414215 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10255060 Supplier Thermo Scientific Acros Supplier No. 120250100
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Edge
This item is not returnable. View return policy

CAS: 73-40-5 | C5H5N5O | 151.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-40-5
Molecular Formula C5H5N5O
Molecular Weight (g/mol) 151.13
MDL Number MFCD00071533
InChI Key UYTPUPDQBNUYGX-UHFFFAOYSA-N
Synonym guanine, 2-amino-6-hydroxypurine, guanin, 2-aminohypoxanthine, mearlmaid, pearl essence, guanine enol, stella polaris, dew pearl, natural pearl essence
PubChem CID 764
ChEBI CHEBI:16235
IUPAC Name 2-amino-3,7-dihydropurin-6-one
SMILES NC1=NC(=O)C2=C(N1)N=CN2

Specifications

Absorbance lambda max. 248nm, pH 1, 1.35 to 1.41 (A250/A260), 0.82 to 0.86 (A280/A260), 0.47 to 0.51 (A290/A260)
Melting Point 360°C
Color Cream to White
Ignition Residue 0.1% max.
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Beilstein 26, 449
Merck Index 15, 4599
Solubility Information Solubility in water: practically insoluble. Other solubilities: freely soluble in alkali hydroxides,dilute acids,sparingly soluble in alcohol and ether
Formula Weight 151.13
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material Guanine
Show More Show Less
compliance-icons
Product Identifier
  • Guanine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.