missing translation for 'onlineSavingsMsg'
Learn More

Guanine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11454543
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11454543

Brand: Thermo Scientific Alfa Aesar A11532.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Guanine hydrochloride is one of the strongest denaturant. It is mainly used in protein purification and protein folding studies. It is a nucleobase used in biochemical research.

Solubility
Slightly soluble in water

Notes
Store in a cool place. Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 635-39-2
Molecular Formula C5H6ClN5O
Molecular Weight (g/mol) 187.59
MDL Number MFCD00213670
InChI Key IBAOFQIOOBQLHE-UHFFFAOYSA-N
Synonym guanine hydrochloride, guanine hcl, 2-amino-1h-purin-6 7h-one hydrochloride, guanine, monohydrochloride, usaf s-1, guanine, hydrochloride, 6h-purin-6-one, 2-amino-1,7-dihydro-, monohydrochloride, 2-amino-9h-purin-6-ol hydrochloride 1:1, 6h-purin-6-one, 2-amino-1,9-dihydro-, hydrochloride 1:1, 2-amino-3,7-dihydropurin-6-one hydrochloride
PubChem CID 69459
IUPAC Name 2-amino-3,7-dihydropurin-6-one;hydrochloride
SMILES Cl.NC1=NC(=O)C2=C(N1)N=CN2

Specifications

Melting Point >300°C
Assay Percent Range 99%
Quantity 250 g
Beilstein 4159901
Merck Index 14,4564
Solubility Information Slightly soluble in water
Formula Weight 187.59
Percent Purity 99%
Chemical Name or Material Guanine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.