missing translation for 'onlineSavingsMsg'
Learn More

Guanine, TRC

Product Code. 30136062
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136062 25 g 25g
17943755 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136062 Supplier Toronto Research Chemicals Supplier No. TRCG83600025G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-40-5
Molecular Formula C5 H5 N5 O
Molecular Weight (g/mol) 151.13
Synonym 2-Amino-1,9-dihydro-6H-purin-6-one, 2-Amino-1,7-dihydro-6H-purin-6-one, Valaciclovir Hydrochloride Anhydrous Imp. A (EP), Ganciclovir Imp. F (EP), Aciclovir Imp. B (EP), Guanine, 6H-Purin-6-one, 2-amino-1,9-dihydro-, 6H-Purin-6-one, 2-amino-1,7-dihydro- (9CI), Guanine (8CI), 2-Amino-6-hydroxy-1H-purine, 2-Amino-6-hydroxypurine, 2-Aminohypoxanthine, 9H-Guanine, C.I. 75170, C.I. Natural White 1, Dew Pearl, Guanin, Guanine enol, Hypoxanthine, 2-amino-, Mearlmaid, Mearlmaid AA, Natural Pearl Essence, Natural White 1, Naturon, Pearl Essence, Stella Polaris, Valganciclovir Hydrochloride Imp. B (EP)
IUPAC Name 2-amino-1,7-dihydropurin-6-one
SMILES NC1=Nc2nc[nH]c2C(=O)N1

Especificaciones

Quantity 25 g
InChI Formula InChI=1S/C5H5N5O/c6-5-9-3-2(4(11)10-5)7-1-8-3/h1H,(H4,6,7,8,9,10,11)
Formula Weight 151.0494
Percent Purity >95%
Chemical Name or Material Guanine
Grade HPLC
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.