missing translation for 'onlineSavingsMsg'
Learn More

Guanosine 5'-Diphosphate Disodium Salt, TRC

Product Code. 30400995
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30400995 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400995 Supplier Toronto Research Chemicals Supplier No. TRCG8379281G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Guanosine 5'-Diphosphate Disodium Salt
CAS 7415-69-2
Molecular Formula C10H13N5Na2O11P2
Quantity 1 g
Synonym Guanosine 5'-(trihydrogen diphosphate), sodium salt (1:2) (ACI), Guanosine 5'-(trihydrogen diphosphate), disodium salt (9CI), Guanosine pyrophosphate, disodium salt (7CI), Guanosine trihydrogen pyrophosphate, disodium salt (8CI), 5'-GDP disodium salt, Disodium guanosine 5'-diphosphate, Guanosine 5'-diphosphate disodium salt
InChI Formula InChI=1S/C10H15N5O11P2.2Na/c11-10-13-7-4(8(18)14-10)12-2-15(7)9-6(17)5(16)3(25-9)1-24-28(22,23)26-27(19,20)21;;/h2-3,5-6,9,16-17H,1H2,(H,22,23)(H2,19,20,21)(H3,11,13,14,18);;/q;2*+1/p-2/t3-,5-,6-,9-;;/m1../s1
SMILES [Na+].[Na+].NC1=Nc2c(ncn2[C@@H]3O[C@H](COP(=O)([O-])OP(=O)(O)[O-])[C@@H](O)[C@H]3O)C(=O)N1
IUPAC Name disodium;[[(2R,3S,4R,5R)-5-(2-amino-6-oxo-1H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] hydrogen phosphate
Molecular Weight (g/mol) 487.16
Formula Weight 486.9882
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.