missing translation for 'onlineSavingsMsg'
Learn More

Guanosine 5'-monophosphate, disodium salt hydrate, 97%

Product Code. 10531671 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10531671 5 g Glass bottle 5g
10618824 25 g Glass bottle 25g
10176633 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10531671 Supplier Thermo Scientific Acros Supplier No. 226600050
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5550-12-9 | C10H12N5Na2O8P | 407.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5550-12-9
Molecular Formula C10H12N5Na2O8P
Molecular Weight (g/mol) 407.19
MDL Number MFCD00151238
InChI Key PVBRXXAAPNGWGE-KHRSEZDTNA-L
PubChem CID 21712
IUPAC Name disodium;[(2R,3S,4R,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES [Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]2O)C(=O)N1

Specifications

Melting Point >300°C
Color White
Loss on Drying 25% max. (120°C, 4 hrs)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Beilstein 10, 279
Merck Index 14, 4570
Synonym guanosine 5'-monophosphate disodium salt, 5'-guanylic acid disodium salt, disodium guanylate, disodium 5'-guanylate, sodium guanylate, disodium gmp, disodium 5'-gmp, gmp disodium salt, 5'-guanylic acid, disodium salt, 5'-gmp disodium salt
Solubility Information Solubility in water: >50g/l water
Spectrophotometry 97% min. (lambda max. 252nm,pH 7) molar absorptivity 13700+/-4%,pH 7
Formula Weight 407.19
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Guanosine 5′-monophosphate,disodium salt hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.