missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Guanosine-5'-triphosphate disodium salt

Product Code. 15414029
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15414029 500 mg 500mg
15404029 250 mg 250mg
15494019 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15414029 Supplier Thermo Scientific Chemicals Supplier No. J61414.ME
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56001-37-7 | C10H14N5Na2O14P3 | 567.14 g/mol

Guanosine 5?-triphosphate (GTP), a purine trinucleotide, is a substrate for RNA polymerases and a wide range of GTPase including G-protein coupled receptor (GPCR) GTPases and cell signaling and cycling associated guanine nucleotide exhange factors (GEF). Guanosine 5?-Triphosphate, Disodium Salt is a phosphoryl donor in protein synthesis and in signal transduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Guanosine 5′-triphosphate (GTP), a purine trinucleotide, is a substrate for RNA polymerases and a wide range of GTPase including G-protein coupled receptor (GPCR) GTPases and cell signaling and cycling associated guanine nucleotide exhange factors (GEF). Guanosine 5′-Triphosphate, Disodium Salt is a phosphoryl donor in protein synthesis and in signal transduction.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56001-37-7
Molecular Formula C10H14N5Na2O14P3
Molecular Weight (g/mol) 567.14
MDL Number MFCD03410297
InChI Key FIZIYLKEXVIRHJ-KHRSEZDTNA-L
Synonym guanosine 5'-triphosphate, guanosine-5'-triphosphate disodium salt dihydrate gtp, guanosine-triphosphate, guanosine 5'-triphosphate 4-, gtp 4-, 2r,3s,4r,5r-5-2-amino-6-hydroxy-9h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxy phosphonatooxy phosphinate, 2r,3s,4r,5r-5-2-amino-6-oxo-3h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl phosphate
PubChem CID 131676145
IUPAC Name disodium;(2R,3R,4S,5R)-2-(2-amino-6-oxo-3H-purin-9-yl)-5-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolane-3,4-diolate
SMILES [Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]2O)C(=O)N1

Specifications

Color White
Quantity 500 mg
Beilstein 4113439
Solubility Information Soluble in water.
Formula Weight 567.15
Physical Form Powder
Chemical Name or Material Guanosine-5'-triphosphate disodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.