missing translation for 'onlineSavingsMsg'
Learn More

Guanosine-5'-triphosphate Disodium Salt (~85%), TRC

Product Code. 30392828
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392828 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392828 Supplier Toronto Research Chemicals Supplier No. TRCG838608250MG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Guanosine-5'-triphosphate Disodium Salt
CAS 56001-37-7
Molecular Formula C10H14N5O14P3Na2
Quantity 250 mg
Synonym Guanosine 5'-(tetrahydrogen triphosphate) sodium salt (1:2), Guanosine 5'-(tetrahydrogen triphosphate) disodium salt (9CI), 5’-GTP di-Na salt, Disodium 5'-GTP, Disodium guanosine triphosphate, GTP disodium salt, Guanosine 5'-triphosphoric acid disodium salt
InChI Formula InChI=1S/C10H16N5O14P3.2Na/c11-10-13-7-4(8(18)14-10)12-2-15(7)9-6(17)5(16)3(27-9)1-26-31(22,23)29-32(24,25)28-30(19,20)21;;/h2-3,5-6,9,16-17H,1H2,(H,22,23)(H,24,25)(H2,19,20,21)(H3,11,13,14,18);;/q;2*+1/p-2/t3-,5-,6-,9-;;/m1../s1
SMILES [Na+].[Na+].NC1=Nc2c(ncn2[C@@H]3O[C@H](COP(=O)(O)OP(=O)(O)OP(=O)([O-])[O-])[C@@H](O)[C@H]3O)C(=O)N1
IUPAC Name disodium;[[[(2R,3S,4R,5R)-5-(2-amino-6-oxo-1H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl] phosphate
Molecular Weight (g/mol) 567.14
Formula Weight 566.9545
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.