missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Guanosine 5'-triphosphate trisodium salt

Product Code. 16461279
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461279 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16461279 Supplier MedChemExpress Supplier No. HY12695
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Guanosine 5'-triphosphate trisodium salt (5'-GTP trisodium salt) is an activator of the signal transducing G proteins which are involved in various cellular processes including proliferation, differentiation, and activation of several intracellular kinase cascades.

Specifications

Molecular Weight (g/mol) 589.13 g/mol
Product Type Natural Products
Content And Storage -20°C, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (stored under nitrogen)
Shipping Condition Room temperature
Purity 98.0%
Quantity 1 g
Solubility Information H2O : ≥ 47 mg/mL (79.78 mM)
CAS 36051-31-7
Synonym 5'-GTP trisodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.