missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Guanosine

Product Code. 16402499
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16402499 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16402499 Supplier MedChemExpress Supplier No. HYN0097
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Guanosine (DL-Guanosine) is a purine nucleoside comprising guanine attached to a ribose (ribofuranose) ring via a β-N9-glycosidic bond. Guanosine possesses anti-HSV activity.

Specifications

Molecular Weight (g/mol) 283.24 g/mol
Product Type Natural Products
Content And Storage 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Shipping Condition Room temperature
Purity 95.01%
Quantity 1 g
Solubility Information DMSO : 100 mg/mL (353.06 mM∣Need ultrasonic) ∣H2O : 1 mg/mL (3.53 mM∣Need ultrasonic)
For Use With (Application) COVID-19-anti-virus
CAS 118-00-3
Synonym DL-Guanosine∣ Vernine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.