missing translation for 'onlineSavingsMsg'
Learn More

Guanosine, 98%

Product Code. 11433734
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
Unit Size:
100g
250g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433734 250 g 250g
11408900 25 g 25g
11423734 50 g 50g
11418900 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11433734 Supplier Thermo Scientific Alfa Aesar Supplier No. A11328.30
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 118-00-3 | C10H13N5O5 | 283.24 g/mol

Guanosine is used as a constituent of nucleic acids. It is used in metallic paints, simulated pearls,plastics,cosmetics industry etc,. It has been also used in pharmacokinetics as a prodrug. Guanosine is used in cell culture applications as a precursor of GMP.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Guanosine is used as a constituent of nucleic acids. It is used in metallic paints, simulated pearls,plastics,cosmetics industry etc,. It has been also used in pharmacokinetics as a prodrug. Guanosine is used in cell culture applications as a precursor of GMP.

Solubility
Soluble in Formic Acid:Water (1:1): 50 mg/mL. Insoluble in methanol, diethyl ether.

Notes
Store away from extreme heat. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-00-3
Molecular Formula C10H13N5O5
Molecular Weight (g/mol) 283.24
MDL Number MFCD00010182
InChI Key NYHBQMYGNKIUIF-UUOKFMHZSA-N
Synonym guanosine, guanine riboside, vernine, guanozin, guanosin, inosine, 2-amino, usaf cb-11, vernine van, l-guanosine, 9-beta-d-ribofuranosylguanine
PubChem CID 6802
ChEBI CHEBI:16750
SMILES NC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1

Specifications

Melting Point ∼250°C (decomposition)
Quantity 250 g
Beilstein 625911
Merck Index 14,4566
Solubility Information Soluble in Formic Acid:Water (1:1): 50mg/mL. Insoluble in methanol,diethyl ether.
Optical Rotation −72° (c=1.5 in 0.1N NaOH)
IUPAC Name 2-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
Formula Weight 283.24
Percent Purity ≥98%
Chemical Name or Material Guanosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.