missing translation for 'onlineSavingsMsg'
Learn More

Gum arabic, powder

Product Code. 10237640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10237640 1 kg Plastic bottle 1kg
10165010 250 g Plastic bottle 250g
10227413 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10237640 Supplier Thermo Scientific Acros Supplier No. 258850010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 9000-01-5

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9000-01-5
MDL Number MFCD00081264
Synonym Acacia

Specifications

Color Beige-Yellow to White
Physical Form Powder
Ash Total ash: < 4.0%
Density 1.3500g/mL
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol
Quantity 1 kg
Identification Passes Test (per Eur. Pharm.)
Infrared Spectrum Authentic
Insoluble Matter 0.1% max.
Loss on Drying 15.0% max. (1 g, 105°C)
Packaging Plastic bottle
Specific Gravity 1.35
Chemical Name or Material Gum arabic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.