missing translation for 'onlineSavingsMsg'
Learn More

Gum Rosin, TRC

Product Code. 30390063
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30390063 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390063 Supplier Toronto Research Chemicals Supplier No. TRCG85550025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gum Rosin
CAS 8050-09-7
Quantity 25 g
Synonym Rosin, Rosin Substitutes, 100F, AAL, AL 104, AL 104 (Rosin), AL 1200, AL 1300, AL 1302, AR 06, AR 06 (Rosin), Aquatac XR 4343, BALS 3A, Bandis G 100, Bandis T 100H, CC 167, CG-WW, CG-X, Chinese Rosin WW, Chinese Rosin X, Colopearl E 5H, Colophane, Colophonium, Colophony, Composize 1718, Composize M 1718, D 101, D 101 (Rosin), DC 35Y01, DS 896, Deep Rosin A 100, Deipurojin A 100, Dermulsene RA 405, Disproportionated Rosin, Dynakoll SI 100, EM 3, EM 3 (rosin), ER 110, Ester Gum EK 1002, FR 1900, Foralyn 5020, G 100F, G 7, GR 90S, Gum Rosin CG-WW, Gum Rosin M, Gum Rosin N, Gum Rosin WW, Gum Rosin X, HBR 509, HRS 50, HRS 50 (Rosin), Hariester 508H, Hariester DS 70, Hariester G 7, Hariester SK 90D55, Hariphenol P 900, Haritac AQ 110, Haritac AQ 90A, Haritac F 90, Harosin X, Harsize L 50, Harsize L 850, Harsize NES 555, Highrosin, Highrosin B, Highrosin S, Hongkong Rosin WW, Inktac 103, Inktac 128, Inktac TX, KA 100W, KE 356, KE 610, KE 709, KR 604, KR 614, KSU 007, Kenomic A 35, Kenomic C 35, L 750, MHK 37BZ, NES 405, NS 74, NT 83, NT 85, Naval stores, Neotall C, NeuSize 3, NeuSize 650, NeuSize 730, NeuSize 737, NeuSize 745, NeuSize 770, Optika, P 90, P 90 (rosin), Pentrex 28, Peroza E 3670, Pexite WG, Pine Resin, Pine Rosin, Pinecrystal KE 356, Pinecrystal KE 614, Pinecrystal KM 5000, Pinecrystal KR 604, Pinecrystal KR 612, Pinecrystal KR 614, Pinecrystal KR 65, Portuguese WW, R 1600, R 515W, RE 802, RF Size 880L50, RF Size E 50A, RR-CHX, RX 0170, Recoldis A, Resarim B 300, Resin 95, Pine Resins, Rosin Resins, Rikatac F 805, Rondis FR-K 15, Rosin WW, SK 501, SN-EX 7337, SN-Tack 750, SPN 771, SPN 775, SPNT 87, Setalin P 7000, Shiragiku Rosin, Shiragiku Rosin 1, Sizepine 775, Solros, Stafor 50, Sylvatac 4100, Sylvatac 96, TAC 3008, TMVS 2N, TSF 25, Unitol HYR, WW 550, WW Rosin, Wood Rosin WG, Wood Rosin, XH 800, YT 102AX, Yellow Resin, YserGum 115, YserGum 116, YserGum 122, YserGum 213
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.