missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GW 0742
GREENER_CHOICE

Product Code. 18014292
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18014292 10 mg 10mg
18075982 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18014292 Supplier Tocris Bioscience™ Supplier No. 2229/10
  • ONLINE EXCLUSIVE PRICE
    328.00 €
    309.75 € / 10mg
    Save 18.25 €
    5% Off
 Restricted Item
Estimated Shipment: 21-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Highly selective, potent PPARδ agonist

Potent and highly selective PPARδ agonist. EC50 values are 0.001, 1.1 and 2 μM for transactivation of human PPARδ, -α, and -γ receptors respectively. Neuroprotective in rat cerebellar granule neuronal cultures after brief (12-hour) exposure but exhibits inherent toxicity after prolonged (48-hour) incubation. Increases rate of fatty acid oxidation and protects against ischemia/reperfusion injury in neonatal and adult cardiomyocytes.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 317318-84-6
Molecular Formula C21H17F4NO3S2

Specifications

Quantity 10 mg
Formula Weight 471.49
Purity >98%
Molecular Weight (g/mol) 471.485
Chemical Name or Material GW 0742
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.