Learn More
Tocris Bioscience™ GW 2580
GREENER_CHOICE
Selective cFMS kinase inhibitor; orally bioavailable
Brand:  Tocris Bioscience™ 5673/10
Encompass
Product Code. 15767036
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
GW 2580 is a potent and selective cFMS kinase inhibitor (IC50 = 0.06 μM). Exhibits selectivity for cFMS kinase over 186 other kinases. Inhibits CSF-1-induced growth of rat monocytes in vitro. Also inhibits bone resorption in osteoclast cultures, as well as connective tissue and bone destruction in a rat adjuvant arthritis model. Suppresses M-NFS-60 tumor cell growth in mice. Orally bioavailable.
Specifications
Specifications
5-[[3-Methoxy-4-[(4-methoxyphenyl)methoxy]phenyl]methyl]-2,4-pyrimidinediamine
870483-87-7