Learn More
DiscoverX GW 4064
Product Code. 17342459
Encompass
Click to view available options
:
10 mg
Product Code. 17342459
Brand: DiscoverX 921129
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
GW 4064 is an agonist for FXR. Provided as 10mg of dry powder.
GW 4064 is a synthetic small molecule which acts as a selective agonist of the orphan nuclear receptor FXR. GW 4064 protects cells and organs against bile acid toxicity, and has been used to elucidate FXR's role in dyslipidemia, diabetes, obesity, and cancer. Has no activity against other nuclear receptors. Provided as 10mg of dry powder.
Specifications
Quantity 10 mg