missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GW 4869

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
GW 4869 is a selective neutral sphingomyelin phosphodiesterase (N-SMase) inhibitor (IC50 = 1 μM). Exhibits selectivity for neutral sphingomyelinase over acid sphingomyelinase (a-SMase) at concentrations up to 150 μM as well as B. cereus PC-PLC, human lyso-PAF PLC, and bovine PP2A at 10 μM. Blocks exosome biogenesis. Inhibits TNF-α-induced sphingomyelin hydrolysis and TNF-α-induced cell death in MCF-7 cells. Reverses hypoxia-induced pulmonary vasoconstriction in rats. Neuroprotective.
Specifications
Specifications
| Chemical Name or Material | 3,3 '-(1,4-Phenylene)bis[N-[4-(4,5-dihydro-1 H-imidazol-2-yl)phenyl]-2-propenamide] ditrifluoroacetate |
| Quantity | 10 mg |
| Target | Hydrolase Inhibitors |
| Molecular Formula | C30H28N6O20.2 CF3COOH |
| Purity | 0.9 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction