missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ GW 9662

Product Code. 18085542
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18085542 10 mg 10mg
18034172 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18085542 Supplier R&D Systems™ Supplier No. 1508/10
  • ONLINE EXCLUSIVE PRICE
    239.00 €
    225.75 € / 10mg
    Save 13.25 €
    5% Off
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective covalent PPARγ antagonist

GW 9662 is a selective irreversible PPARγ antagonist (IC50 values are 3.3, 32 and 2000 nM for PPARγ, PPARα and PPARδ respectively). Blocks the inhibition of osteoclast formation induced by IL-4 in the low micromolar range (1-2 μM), therefore is more potent than BADGE. Anticancer, inhibits growth of human mammary tumor cell lines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22978-25-2
Molecular Formula C13H9ClN2O3

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 276.68
Purity >98%
Molecular Weight (g/mol) 276.676
Chemical Name or Material GW 9662
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.