missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies GW2580

Product Code. 17140072
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17140072 25 mg 25mg
17130072 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17140072 Supplier STEMCELL Technologies Supplier No. 72474
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

CSF-1 pathway inhibitor; Inhibits cFMS

  • GW2580 is a selective inhibitor of cFMS kinase (IC50 = 0.03 μM) (Conway et al.), 2008), blocking its ability to autophosphorylate colony-stimulating factor (CSF-1 or M-CSF), which promotes the survival, proliferation, and differentiation of macrophages
  • Demonstrates the importance of CSF-1 in promoting myeloid lineage bias in mouse hematopoietic stem cells (Mossadegh-Keller et al.)
  • Demonstrates the importance of CSF-1 in expansion of mouse and human macrophage colonies and monocytes (He et al.; Clanchy et al.; Conway et al.), 2008; Conway et al.), 2005)
  • Inhibits bone degradation in cultures of human osteoclasts, rat calvaria, and rat fetal long bone (Conway et al.), 2005), and in mouse models of arthritis (Conway et al.), 2008)

Specifications

CAS 870483-87-7
Quantity 25 mg
Molecular Formula C20H22N4O3
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.