missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ H 1152 dihydrochloride
GREENER_CHOICE

Product Code. 30001317
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001317 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001317 Supplier Tocris Bioscience™ Supplier No. 2414/1
  • ONLINE EXCLUSIVE PRICE
    341.00 €
    322.35 € / 1mg
    Save 18.65 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Selective Rho-kinase (ROCK) inhibitor

Rho-kinase (ROCK) inhibitor that displays high selectivity over other protein kinases (IC50 values are 0.012, 0.180, 0.360, 0.745, 3.03, 5.68 and 28.3 μM for ROCKII, CAMKII, PKG, Aurora A, PKA, PKC and MLCK respectively). Inhibits sulprostone-induced contractions in guinea pig aorta (IC50 = 190 nM) and displays proerectile effects in rats.

Glycyl Derivative also available.

TRUSTED_SUSTAINABILITY

Specifications

CAS 871543-07-6
Quantity 1 mg
Molecular Formula C16H23Cl2N3O2S
Formula Weight 392.34
Purity >98%
Molecular Weight (g/mol) 392.339
Chemical Name or Material H 1152 dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.