Learn More
Bel-Art™ H-B Durac Plus Ultra Low Liquid-In-Glass Laboratory Thermometer
Bel-Art, H-B Durac Plus Liquid-in-Glass Laboratory Thermometers are partial and total immersion thermometers for Low Temperature Applications.
Brand: Bel-Art™ B60760-1200
Additional Details : Weight : 0.03720kg Transport : UN number : 1294 Chem class : 3 Pack group : II
Description
- Designed for ultra-low temperature use to ensure that the meniscus will be visible in the appropriate low temperature applications, these thermometers feature mercury free liquid against lead-free, white back glass.
- Traceable to NIST
- Include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability
- Tested and inspected by our USA quality team
- For enhanced readability use thermometer magnifier B61400-7000
- Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.
Specifications
Low Temperature Applications | |
Statement of Accuracy and Traceability | |
Thermometer | |
-100°C | |
76 mm |
Total or Partial | |
Glass | |
50°C | |
-100°C to +50°C |