Learn More
Thermo Scientific™ H-Flex 1 Swinging Bucket Rotor and Accessories
Spin more with less! High capacity and flexibility—all in a small footprint.
137.00 € - 2895.00 €
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
16525311
|
Thermo Scientific™
75003303 |
Thermo Scientific H-Flex 1 Rotor 8x50mL Adapter, set/2 |
840.00 €
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16535311
|
Thermo Scientific™
75003304 |
Thermo Scientific H-Flex 1 Rotor 18x15mL Adapter, set/2 |
840.00 €
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16545311
|
Thermo Scientific™
75003305 |
Thermo Scientific H-Flex 1 Rotor 33x10/12mL Adapter, set/2 |
835.00 €
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16555311
|
Thermo Scientific™
75003306 |
Thermo Scientific H-Flex 1 Rotor 42x5/7mL Adapter, set/2 |
835.00 €
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16575311
|
Thermo Scientific™
75003308 |
Thermo Scientific H-Flex 1 Rotor 400mL Bottle/Centrilab Adapter, set/2 |
835.00 €
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16595301
|
Thermo Scientific™
75003300 |
Thermo Scientific H-Flex 1 Rotor Cross |
2895.00 €
Each |
Estimated Shipment: 16-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16505311
|
Thermo Scientific™
75003301 |
Thermo Scientific H-Flex 1 Rotor Buckets, set/2 |
1830.00 €
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16515311
|
Thermo Scientific™
75003302 |
Thermo Scientific H-Flex 1 Rotor Biocontainment Lids, set/2 |
411.00 €
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16565311
|
Thermo Scientific™
75003307 |
Thermo Scientific H-Flex 1 Rotor Microplate Adapter, set/2 |
835.00 €
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16585311
|
Thermo Scientific™
75003309 |
Thermo Scientific H-Flex 1 Rotor Replacement O-Rings, set/2 |
137.00 €
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Spin more with less! Whether clinical and diagnostic applications or cell culture and other research, now spin conical tubes, blood tubes and microplates all in one rotor for a small-footprint General Purpose centrifuge. High capacity and flexibility—all in a small footprint.
- Auto-Lock and Click Seal biocontainment lids enable fast, one-handed rotor exchanges and lid locking
- Spins blood tubes, conical tubes and microplates without changing rotor or buckets
- Get results faster without the need to change hardware
- Limit rotor maintenance and contamination potential with slide-coat technology eliminating the need for grease and lubricants
Specifications
Swinging Bucket Rotor |