missing translation for 'onlineSavingsMsg'
Learn More

H-Glu-Lys-Oh, TRC

Product Code. 30240791
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240791 100 mg 100mg
30240586 50 mg 50mg
30240509 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240791 Supplier Toronto Research Chemicals Supplier No. TRCG650303100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5891-53-2
Molecular Formula C11H21N3O5
Molecular Weight (g/mol) 275.31
Synonym L-alpha-Glutamyl-L-lysine, 179: PN: WO2011146121 PAGE: 117 Claimed Sequence, 459: PN: WO2005081628 SEQID: 461 Claimed Protein, 7: PN: WO2018204847 SEQID: 7 Claimed Protein, Glutamyllysine, N2-L-alpha-Glutamyllysine, alpha-Glu-Lys
IUPAC Name (2S)-6-amino-2-[[(2S)-2-amino-4-carboxybutanoyl]amino]hexanoic acid
SMILES NCCCC[C@H](NC(=O)[C@@H](N)CCC(=O)O)C(=O)O

Specifications

InChI Formula InChI=1S/C11H21N3O5/c12-6-2-1-3-8(11(18)19)14-10(17)7(13)4-5-9(15)16/h7-8H,1-6,12-13H2,(H,14,17)(H,15,16)(H,18,19)/t7-,8-/m0/s1
Quantity 100 mg
Formula Weight 275.1481
Shipping Condition Room Temperature
Chemical Name or Material H-Glu-Lys-Oh
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.