missing translation for 'onlineSavingsMsg'
Learn More

H-Lys(biotinyl)-OH, 98%, BLD Pharm™

Product Code. 31100512
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31100512 100 mg
31103751 5 g
31094339 250 mg
31105692 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31100512 Supplier BLD Pharm Supplier No. BD116725100mg
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 576-19-2
Molecular Formula C16H28N4O4S
MDL Number MFCD00077319
Quantity 100 mg
SMILES O=C(O)[C@@H](N)CCCCNC(CCCC[C@@H]1SC[C@]([C@]1([H])N2)([H])NC2=O)=O
Molecular Weight (g/mol) 372.48
Percent Purity 98%
Chemical Name or Material H-Lys(biotinyl)-OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.