missing translation for 'onlineSavingsMsg'
Learn More

HA14-1, MedChemExpress

Product Code. 16494908
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16494908

Brand: Medchem Express HY12011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HA14-1 is a Bcl-2/Bcl-XL antagonist. HA14-1 binds the designated pocket on Bcl-2 with the IC50 of ≈9 μM in competing with the Bcl-2 binding of Flu-BakBH3, and inhibits its function.

HA14-1 is a Bcl-2/Bcl-XL antagonist. HA14-1 binds the designated pocket on Bcl-2 with the IC50 of ≈9 μM in competing with the Bcl-2 binding of Flu-BakBH3, and inhibits its function.

Specifications

Chemical Name or Material HA14-1
CAS 65673-63-4
Color White
Purity Grade Notes Research
Molecular Formula C17H17BrN2O5
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (122.18 mM)
SMILES CCOC(C1=C(N)OC2=CC=C(Br)C=C2[C@@]1([H])[C@H](C#N)C(OCC)=O)=O
Molecular Weight (g/mol) 409.23
Formula Weight 409.23
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.