Learn More
IKA™ HABITAT Cell, Double-walled 0.5 L Package
Product Code. 17705079 Shop All IKA Products
Change view
Click to view available options
Material:
Borosilicate Glass
Unit Size:
Each
Product Code. Material Unit Size
17705079 Borosilicate Glass Each
1 options
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 17705079 Supplier IKA™ Supplier No. 0010007645
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
The HABITAT cell package for the bioreactor HABITAT research contains a 0.5 L double-walled glass vessel for cell culture applications as well as the appropriate stirring drive.
Together with the separately available control tower package (e.g. HABITAT cell) you receive all necessary components for a successful cultivation. For temperature control via the double jacket we recommend one of our our circulators such as the HRC basic or control.
Scope of delivery
  • Stand 0.5
  • HA.gv.dw.0.5 Glass vessel, double-wall
  • HA.mt.s.2 Motor, small
  • HA.hp.s.0.5 Harvest pipe, straight
  • HA.fd Feeding port (2 pcs.)
  • HA.sp.m.0.5 Micro sparger, 5 μm
  • HA.cn Condenser
  • HA.ip.pi.0.5 3-pitched blade impeller
  • HA.s.tm.1 Temperature sensor
  • HA.s.ph.0.5 pH sensor
  • HA.s.do.0.5 DO sensor
  • HA.s.fo Foam sensor
  • HA.s.lv.1 Level sensor
  • HA.cab.dw Cable and tube set
  • HA.sf.250 Sample flask (4 pcs.)
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 0.5 L
Material Borosilicate Glass