Learn More
Hafnium(IV) chloride, 98+% (metals basis excluding Zr), Zr <2.7%
Description
Hafnium chloride tetrahydrofuran complex is an efficient catalyst for the Diels-Alder cycloadditions, Ziegler-Natta polymerization of alkenes. It is used as control rods in nuclear reactors and in manufacture of light bulb filaments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 320° (sublimation) |
| Color | White |
| Physical Form | Powder |
| Quantity | 2.5 g |
| Assay Percent Range | >98% (Metals basis excluding Zr) |
| Solubility Information | Soluble in water,methanol and acetone. |
| Formula Weight | 320.3 |
| Odor | Irritating |
| Sensitivity | Moisture sensitive |
| Chemical Name or Material | Hafnium(IV) chloride |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.