missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Halofuginone hydrobromide
Description
Halofuginone hydrobromide is a high affinity competitive prolyl-tRNA synthetase inhibitor (Ki = 18.3 nM). Blocks expression of MMP2. Inhibits ECM invasion in vitro and lung metastasis by bladder cancer cells in mice. Inhibits the development of Th17-driven autoimmunity in a mouse model of multiple sclerosis by activating the amino acid response (AAR) pathway. Also antiparasitic.
Specifications
Specifications
| Chemical Name or Material | (2R*,3S*)-7-Bromo-6-chloro-3-[3-(3-hydroxy-2-piperidinyl)-2-oxopropyl]-4-3H-quinazolinone hydrobromide |
| CAS | 64924-67-0 |
| Assay Percent Range | >98% |
| Quantity | 50 mg |
| Molecular Formula | C16H17BrClN3O3.HBr |
| Formula Weight | Observed MW: 495.59 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?