missing translation for 'onlineSavingsMsg'
Learn More

Haloperidol Nonanoate, TRC

Product Code. 30133092
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133092 10 mg 10mg
30012857 100 mg 100mg
30007655 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133092 Supplier Toronto Research Chemicals Supplier No. TRCH10374010MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2512216-30-5
Molecular Formula C30H39ClFNO3
Molecular Weight (g/mol) 516.09
IUPAC Name 4-(4-chlorophenyl)-1-(4-(4-fluorophenyl)-4-oxobutyl)piperidin-4-yl nonanoate
SMILES O=C(CCCN1CCC(C2=CC=C(C=C2)Cl)(CC1)OC(CCCCCCCC)=O)C3=CC=C(C=C3)F

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C30H39ClFNO3/c1-2-3-4-5-6-7-10-29(35)36-30(25-13-15-26(31)16-14-25)19-22-33(23-20-30)21-8-9-28(34)24-11-17-27(32)18-12-24/h11-18H,2-10,19-23H2,1H3
Formula Weight 515.26
Chemical Name or Material Haloperidol Nonanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.