Learn More
Thermo Scientific™ Halt™ Protease and Phosphatase Inhibitor Cocktail, EDTA-free (100X)
Shop All Thermo Scientific ProductsDescription
The all-in-one Thermo Scientific™ Halt Protease and Phosphatase Inhibitor Cocktail provides the convenience of a single solution with full sample protection.
This broad-spectrum Halt Protease and Phosphatase Inhibitor Cocktail is specifically optimized to protect proteins from degradation during extraction and purification. The cocktail contains inhibitors against the major classes of proteases and phosphatases, targeting aminopeptidases, cysteine and serine proteases and serine/threonine and protein tyrosine phosphatases. The 100X cocktails, available in regular and EDTA-free formulations and several package formats, are refrigerator-stable liquids that are very easy to use for nearly any sample or experimental scale.
Highlights:
- Complete protection – all-in-one cocktail contains both protease and phosphatase inhibitors
- Multiple package sizes – choose from four convenient packages sizes to match nearly any scale
- Convenient – the 100X liquid formulation is much easier and more effective to use than individual inhibitors and other formats
- Easy storage – aqueous-based format allows for convenient cold-room storage without freezing, by contrast with DMSO-based cocktails
- Compatible – can be used in mass spectrometry (MS) because it does not contain AEBSF, which can cause peaks to shift
- Validated – fully compatible with Thermo Scientific Pierce™ Cell Lysis Buffers
Includes:
Formulations with and without supplementary vial of EDTA for metalloprotease inhibition, both of which are offered in four package sizes.
Storage Requirements:
Upon receipt store at 4°C. Do not freeze product.
Specifications
Specifications
| Description | Halt Protease and Phosphatase Inhibitor Cocktail, EDTA-Free |
| Product Type | Protease and Phosphatase Inhibitor Cocktail |
| Content And Storage | Upon receipt store at 4°C. Product shipped with an ice pack. Do not freeze product. |
| Form | Liquid |
| Product Line | Halt |
| Buffer | Lysis Buffer |
| Reagent Type | Protease Inhibitor, Phosphatase Inhibitor |
| Inhibitors | Protease Inhibitor, Phosphatase Inhibitor |
| Quantity | 10 mL |
Safety and Handling
- Protease and Phosphatase Inhibitor Cocktail
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- MIXTURE LIST-Contain: Sodium fluoride, Trisodium orthovanadate
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.