Learn More
Hamilton™ 1000 Series Gastight™ Syringes: Luer Lock Syringes, TLL Termination
Mid-volume GASTIGHT syringe ideal for dispensing volumes from 100 μL up to 100 mL.
98.85 € - 534.00 €
Specifications
For Use With (Application) | Ideal for manual and automated HPLC injections, commonly used in syringe pumps and other liquid handling instruments |
---|---|
Product Type | Gastight Syringe |
Needle Point Number | No Needle Included |
Quantity | Ea. |
Product Code | Brand | Volume (Metric) | Model | Connection Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Model | Connection Type | Price | Quantity & Availability | |||
10611353
|
Hamilton™
85027 |
50 mL | 1050 | Luer Lock with Slots |
375.00 €
Each |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10235672
|
Hamilton™
82527 |
25 mL | 1025 | Luer Lock with Slots |
398.00 €
Each |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10040092
|
Hamilton™
86020 |
100 mL | 1100 | Luer Lock |
534.00 €
Each |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10772361
|
Hamilton™
81320 |
1 mL | 1001 | Luer Lock |
98.85 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10120031
|
Hamilton™
81420 |
2.5 mL | 1002 | Luer Lock |
101.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10500052
|
Hamilton™
81520 |
5 mL | 1005 | Luer Lock |
112.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10683921
|
Hamilton™
81620 |
10 mL | 1010 | Luer Lock |
128.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10374902
|
Hamilton™
81327 |
1 mL | 1001 | Luer Lock with Slots |
110.00 €
Each |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10531213
|
Hamilton™
81427 |
2.5 mL | 1002 | Luer Lock with Slots |
103.00 €
Each |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10757484
|
Hamilton™
81527 |
5 mL | 1005 | Luer Lock with Slots |
130.00 €
Each |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10334812
|
Hamilton™
81627 |
10 mL | 1010 | Luer Lock with Slots |
206.00 €
Each |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10495762
|
Hamilton™
82520 |
25 mL | 1025 | Luer Lock |
368.00 €
Each |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10674301
|
Hamilton™
85020 |
50 mL | 1050 | Luer Lock |
460.00 €
Each |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Mid-volume GASTIGHT syringe ideal for dispensing volumes from 100 μL up to 100 mL.
- PTFE Luer Lock (TLL)- This termination has a PTFE, male Luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors
- The TLL is used with Hamilton diluters/dispensers, OEM applications, and manual operations
- The version with slots is used during the attachment to some instruments but does not effect the connection to needles
- Autoclavable when disassembled, except on 25mL and greater syringes; repeated autoclaving will shorten syringe life
- Needle point styles 4 and AS are available on request)
Specifications
Ideal for manual and automated HPLC injections, commonly used in syringe pumps and other liquid handling instruments | |
Gastight Syringe | |
No Needle Included | |
Ea. |