Learn More
Hamilton™ 1800 Series Gastight™ Replacement Syringe Barrels
Product Code. 15573963
Change view
Click to view available options
For Use With (Equipment):
1810 RN Syringe
Volume (Metric):
100 μL
Unit Size:
Each
Product Code. For Use With (Equipment) Volume (Metric) unitSize
15573963 1810 RN Syringe 100 μL Each
1 options
Product Code. 15573963 Supplier Hamilton™ Supplier No. 32190
Add to basket
Add to basket
Barrels range in volume from 10μ to 250μL with removable needle (RN) and cemented needle (N) terminations.
Specifications
Material PTFE
For Use With (Application) Gases and liquids
Product Type Replacement Syringe Barrel
Volume (Metric) 100 μL
For Use With (Equipment) 1810 RN Syringe