Learn More
Hamilton™ Disposable Tip Adapter
Product Code. 10065610
Change view
Click to view available options
Volume (Metric):
100 μL
Unit Size:
Each
Product Code. Volume (Metric) Unit Size
10065610 100 μL Each
1 options
Product Code. 10065610 Supplier Hamilton™ Supplier No. 31330
  • ONLINE EXCLUSIVE PRICE
    43.75 €
    39.32 € / Pack of 1
    Save 4.43 €
    10% Off
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed to enable the connection of a standard 1mL disposable pipette tip to the end of a Hamilton™ syringe.
  • Adapter connects to standard Luer Tip (LT) and PTFE Luer Lock (TLL) syringes
  • Ideal for eliminating the possibility for cross-contamination
  • Manufactured from CTFE
  • Provided individually
Specifications
Material CTFE
Product Type Luer-tip Adapter
Volume (Metric) 100 μL
For Use With (Equipment) PB600-1 Repeating Dispensers