Learn More
Hamilton™ Microlab Accessories - Printer Kit
Click to view available options
:
Microlab Printer Kit
Description
- Satisfies regulatory CFR21 Part 11 and FDA GMP/GLP requirements by conveniently printing all log files directly from the device's controller
- Microlab 600 diluter/dispenser printer kit interfaces with the free compliance software package that is available for Advanced devices
- Kit automatically is compatible with all Microlab 300 pipettors
- Prints on standard, non-thermal 8.5x11 and A4 paper
Specifications
Specifications
| For Use With (Equipment) | Microlab™ 600 diluter, Microlab™ 300 pipetteting system |
| Includes | Printer, power source, adapter and cable, free compliance software package |