Learn More
Hamilton™ RN Adapters (Removable Adapters)
Click to view available options
Type:
Female Luer-lock Hub Adapter
Male Luer-lok Hub Adapter
Unit Size:
Each
Description
Adapters provide a connection to threaded fittings, tubing assemblies, tees, and crosses.
Specifications
Specifications
| Type | Female Luer-lock Hub Adapter |
| For Use With | Removable Needle Syringe (250μL and Larger) |