missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Small Hub Removable Needles (2.5 - 100μL Syringes)

Can be used with removable (RN)-style syringes that are 100μL and smaller.

Brand:  Hamilton™ 7762-02

Code : A0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11500142

  • 193.00 € / Pack of 6
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Standard needle length is 51mm (2 in.)
  • Custom gauges, lengths and point styles are available
Specifications

Specifications

Autoclavable
28 G
0.014 in.
0.36 mm
2 in.
Blunt
Stainless Steel (Hub)
0.007 in.
0.18 mm
Ideal when there is a risk of the needle clogging
Replacement Syringe Needle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers