Learn More
Hamilton™ Small Hub Removable Needles (2.5 - 100μL Syringes)
Can be used with removable (RN)-style syringes that are 100μL and smaller.
52.00 € - 269.00 €
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Stainless Steel (Hub) |
Length (English) Needle | 2 in. |
Product Type | Replacement Syringe Needle |
Product Code | Brand | Needle Gauge | Diameter (English) Inner | Diameter (English) Outer | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Needle Gauge | Diameter (English) Inner | Diameter (English) Outer | Price | Quantity & Availability | |||
10291733
|
Hamilton™
7784-07 |
26s G | 0.0051 in. | 0.018 in. |
204.00 €
Pack of 6 |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10708494
|
Hamilton™
7784-08 |
26 G | 0.01 in. | 0.018 in. |
204.00 €
Pack of 6 |
Estimated Shipment: 23-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10284302
|
Hamilton™
7784-06 |
22 G | 0.016 in. | 0.028 in. |
204.00 €
Pack of 6 |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10622804
|
Hamilton™
7758-01 |
22 G | 0.016 in. | 0.028 in. |
54.00 €
Pack of 6 |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10274702
|
Hamilton™
7784-05 |
22s G | 0.0059 in. | 0.028 in. |
204.00 €
Pack of 6 |
Estimated Shipment: 23-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10323811
|
Hamilton™
7758-03 |
22s G | 0.0059 in. | 0.028 in. |
54.00 €
Pack of 6 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11557704
|
Hamilton™
7770-01 |
22s G | 0.0059 in. | 0.028 in. |
66.00 €
Pack of 6 |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11520142
|
Hamilton™
7762-06 |
33 G | 0.004 in. | 0.008 in. |
178.00 €
Pack of 6 |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10152642
|
Hamilton™
7770-02 |
22 G | 0.016 in. | 0.028 in. |
69.00 €
Pack of 6 |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10225142
|
Hamilton™
7768-01 |
26s G | 0.0051 in. | 0.018 in. |
67.00 €
Pack of 6 |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10788731
|
Hamilton™
7758-02 |
26s G | 0.0051 in. | 0.019 in. |
52.00 €
Pack of 6 |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11547704
|
Hamilton™
7768-02 |
26 G | 0.01 in. | 0.01 in. |
67.00 €
Pack of 6 |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11503234
|
Hamilton™
7758-04 |
26 G | 0.01 in. | 0.018 in. |
53.00 €
Pack of 6 |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10616264
|
Hamilton™
7762-03 |
30 G | 0.0063 in. | 0.012 in. |
181.00 €
Pack of 6 |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11510142
|
Hamilton™
7762-05 |
32 G | 0.004 in. | 0.0094 in. |
173.00 €
Pack of 6 |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10183352
|
Hamilton™
7762-04 |
31 G | 0.005 in. | 0.01 in. |
173.00 €
Pack of 6 |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11500142
|
Hamilton™
7762-02 |
28 G | 0.007 in. | 0.014 in. |
193.00 €
Pack of 6 |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10281293
|
Hamilton™
7762-01 |
27 G | 0.0051 in. | 0.016 in. |
269.00 €
Pack of 6 |
Estimated Shipment: 23-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Standard needle length is 51mm (2 in.)
- Custom gauges, lengths and point styles are available
Specifications
Autoclavable | |
Stainless Steel (Hub) | |
2 in. | |
Replacement Syringe Needle |