Learn More
Hamilton™ Syringe Guide for Microliter™ and Gaslight™ Syringes
Product Code. 10130221
Change view
Click to view available options
For Use With (Equipment):
Microliter, Gastight and 700 and 1700 Series Syringes
Microliter, Gastight, 7000 Series and 25 to 500μL Syringes
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10130221 Microliter, Gastight and 700 and 1700 Series Syringes Each
10606473 Microliter, Gastight, 7000 Series and 25 to 500μL Syringes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10130221 Supplier Hamilton™ Supplier No. 14806
  • ONLINE EXCLUSIVE PRICE
    60.80 €
    54.62 € / Pack of 1
    Save 6.18 €
    10% Off
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed to work with 700/7000 series MICROLITER syringes and 1700 series GASTIGHT syringes
Syringe guides are designed to prevent plunger bending and the adjustable stop prevents plunger damage.
Specifications
Product Type Syringe Guide
For Use With (Equipment) Microliter, Gastight and 700 and 1700 Series Syringes